Ethylhippurat ≥98%
Lieferant: Thermo Scientific
B24765.14EA
215
EUR
B24765.14
B24765.06
Ethylhippurat ≥98%
Ethylhippurat
Formel:
C₁₁H₁₃NO₃ Molecular Weight: 207,23 g/mol Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026890 CAS-Nummer: 1499-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...