N-Boc-L-tyrosin

Lieferant: Molekula

MOLE31623559-25GEA 68.7 EUR
MOLE31623559-25G
N-Boc-L-tyrosin
N-Boc-L-tyrosin
Formel: C₁₄H₁₉NO₅
Molecular Weight: 281,3 g/mol
MDL: MFCD00037179
CAS-Nummer: 3978-80-1
EINECS: 223-613-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR