3-Amino-2-methyl-5-nitrobenzamid, Supelco®

Lieferant: Merck

32731-10MGEA 360 EUR
32731-10MG
3-Amino-2-methyl-5-nitrobenzamid, Supelco®
3-Amino-2-methyl-5-nitrobenzamid
Formel: C₈H₉N₃O₃
MDL: MFCD01679715
CAS-Nummer: 3572-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR