DL-Phenylalaninol

Lieferant: ABCR GMBH

ABCRAB153943.0005EA 0 EUR
ABCRAB153943.0005
DL-Phenylalaninol
DL-Phenylalaninol
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
MDL: MFCD00066689
CAS-Nummer: 16088-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR