(Chlormethyl)dimethylpentylsilan

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO009686-10GEA 0 EUR
FLUO009686-10G SILNSILS04035.5 FLUO009686-25G SILNSILS04035.1
(Chlormethyl)dimethylpentylsilan
(Chlormethyl)dimethylpentylsilan
Formel: C₈H₁₉ClSi
MDL: MFCD00674506
CAS-Nummer: 73013-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR