1-(3-(Trifluormethyl)phenyl)thioharnstoff, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL632937-5GEA 52.8 EUR
SIAL632937-5G
1-(3-(Trifluormethyl)phenyl)thioharnstoff, Sigma-Aldrich®
1-(3-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
MDL: MFCD00041171
CAS-Nummer: 1736-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR