5-Methylpicolinsäure

Lieferant: BEPHARM LTD.

BEPHB104894EA 0 EUR
BEPHB104894
5-Methylpicolinsäure
5-Methylpicolinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137,1360 g/mol
MDL: MFCD01704358
CAS-Nummer: 4434-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR