1,4-Oxazepan Hydrochlorid

Lieferant: SIA ENAMINE

ENAMEN300101939.1GEA 0 EUR
ENAMEN300101939.5G ENAMEN300101939.1G ENAMEN300101939
1,4-Oxazepan Hydrochlorid
1,4-Oxazepan Hydrochlorid
Formel: C₅H₁₂ClNO
Molecular Weight: 137,61 g/mol
MDL: MFCD02683070
CAS-Nummer: 5638-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR