Diethoxymethylsilan, Sigma-Aldrich®

Lieferant: Merck

SIAL66612-10MLEA 78.8 EUR
SIAL66612-10ML
Diethoxymethylsilan, Sigma-Aldrich®
Diethoxymethylsilan
Formel: C₅H₁₄O₂Si
Molecular Weight: 134,25 g/mol
MDL: MFCD00026749
CAS-Nummer: 2031-62-1
EINECS: 217-982-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR