Diethoxymethylvinylsilan

Lieferant: CHEMPUR

CHMPSILS06262.EA 0 EUR
CHMPSILS06262.
Diethoxymethylvinylsilan
Diethoxymethylvinylsilan
Formel: C₇H₁₆O₂Si
Molecular Weight: 160,29 g/mol
MDL: MFCD00009067
CAS-Nummer: 5507-44-8
EINECS: 226-850-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR