5-(Trifluormethyl)picolinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL700630-500MGEA 131 EUR
SIAL700630-500MG
5-(Trifluormethyl)picolinsäure, Sigma-Aldrich®
5-(Trifluormethyl)picolinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
MDL: MFCD04113632
CAS-Nummer: 80194-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR