Kaliumethyltrifluoroborat, Sigma-Aldrich®

Lieferant: Merck

SIAL715212-1GEA 83.6 EUR
SIAL715212-1G
Kaliumethyltrifluoroborat, Sigma-Aldrich®
Potassium ethyltrifluoroborate
Formel: C₂H₅BF₃K
Molecular Weight: 135,9656 g/mol
MDL: MFCD04112713
CAS-Nummer: 44248-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR