Dimethylammoniumchlorid

Lieferant: LGC Standards PROMOCHEM
Achtung

Synonyme: Dimethylamin Hydrochlorid

PROOMM0056.06EA 515 EUR
PROOMM0056.06
Dimethylammoniumchlorid
Dimethylammoniumchlorid
Formel: C₂H₈ClN
Molecular Weight: 81,54 g/mol
Schmelzpunkt: 170…173 °C
MDL: MFCD00012477
CAS-Nummer: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR