Wasser ACS, Reagenzqualität ASTM Type I, ASTM Type II Packaged in plastic containers
Lieferant: Ricca Chemical
Synonyme:
Wasserstoffoxid
, Hydrogenoxid
RICC9150-25EA
0
EUR
RICC9150-25
RICC9150000-20F
Wasser ACS, Reagenzqualität ASTM Type I, ASTM Type II Packaged in plastic containers
Wasser
This product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, the specifications below at the time of packaging. This water also meets ASTM Type III and Type IV specifications.
Formel:
H₂O Molecular Weight: 18,02 g/mol Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Appearance | Colorless liquid |
Chloride (Cl) | max 1 ppb |
Conductivity at 25°C (Method: SQCP031) | max 0.056 µS/cm |
Heavy Metals (as Pb) | max 0.01 ppm |
Nitrate (NO₃) | max 0.4 ppm |
Organic Carbon (TOC) | max 50 ppb |
Oxidizable Substances (Permanganate Retention) | To Pass Test |
Phosphate (PO₄) | max 1.0 ppm |
Silicate (as SiO₂) | max 3 ppb |
Sodium (Na) | max 1 ppb |
Sulfate (SO₄) | max 1.0 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...