2-(Methoxycarbonyl)phenylboronsäure

Lieferant: BEPHARM LTD.

BEPHB6448-100GEA 0 EUR
BEPHB6448-100G
2-(Methoxycarbonyl)phenylboronsäure
2-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,97 g/mol
MDL: MFCD02179452
CAS-Nummer: 374538-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR