4-Brom-1-fluor-2-iodbenzol

Lieferant: BEPHARM LTD.

BEPHZB9749-100GEA 0 EUR
BEPHZB9749-100G
4-Brom-1-fluor-2-iodbenzol
4-Brom-1-fluor-2-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9000 g/mol
MDL: MFCD07368751
CAS-Nummer: 116272-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR