Beta Radiation Waste Boxes
Lieferant: Köhler Technische Produkte
KOEHA-1123EA 659 EUR
KOEHA-1123
Beta Radiation Waste Boxes
Boxen Beta-/Gammastrahlen-Sicherheitsbehälter
Beta radiation waste disposal boxes, acrylic, with tightly closing hinged lid.
  • Suitable for disposal of other critical materials
  • Lid provides protection against contaminated materials
Material thickness: 10 mm