Terry tool tubing cutters
Lieferant: Thermo Fisher Scientific
HYPE60182-510EA 438 EUR
HYPE60182-510
Terry tool tubing cutters
Schneidewerkzeuge
Tubing cutter cuts ¹/₈" stainless steel tubing.
Produce clean, 90° cuts of ¹/₈" stainless steel tubing.