Fingertight adapters, Color-It
Lieferant: VICI
Color-It
JOURJR-55016-12EA 21.2 EUR
JOURJR-55016-12 JOURJR-55014-5 JOURJR-55015-5 JOURJR-55012-5 JOURJR-55013-5 JOURJR-55010-5 JOURJR-55011-5
Fingertight adapters, Color-It
Fittings und Verbinder
These fingertight PP adapters are used with ¹/₄" PEEK and stainless hex-head nuts, converting them into colour coded fingertight fittings.
  • Fit all hex-head PEEK and SS fittings for ¹/₁₆" tubing
  • Used in low and high pressure
  • Available in six colours for easy solvent line identification