Waste container, SHARPSGUARD® eco pharmi
Lieferant: DANIELS
Sharpsguard®
DANIDD205EA 204 EUR
DANIDD220_U DANIDD205 DANIDD210
Waste container, SHARPSGUARD® eco pharmi
Behälter Abfallbehälter
SHARPSGUARD® eco pharmi are ideal general-purpose waste containers for the disposal of pharmaceutical waste in original packaging, such as unused pills, no sharps.
  • Aperture in the lid which can be closed temporarily to prevent accidental spillage
  • Sustainable: Base, lid and strap are made from 100% recycled plastic
  • With handle
These containers are for disposal by incineration or other authorised process and are designed for the consignment of waste categorised by EWC codes: 18-01-09 & 18-02-08.