Swivel pad holder, Perfex
Lieferant: Perfex
129-0120EA 71.3 EUR
129-0120 129-0185
Swivel pad holder, Perfex
Mopps
Swivel pad holder is designed to twist and turn for cleaning hard-to-reach areas. Its velcro base holds pads securely in place. Attach a polymer adjustable handle to the pad holder to clean floors, baseboards, walls, windows, and ceilings. Also, ideal for cleaning fine surfaces, including sinks, vinyl, and stainless steel.
Bestellinformation: pads, and handle are sold separately, please contact us for details.