Protein A, HRP (Horseradish Peroxidase)
Lieferant: Cytiva
NA9120-1MLEA 591 EUR
GEHE25-0051-72 NA9120-1ML
Protein A, HRP (Horseradish Peroxidase)
Protein- Expression Proteinexpressions-Reagenzien
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species. For use in immunodetection and in situ hybridisation applications.