pH sensor, InPro3253/SG/120/PT1000
Lieferant: Mettler-Toledo
InPro®
METN52002577EA 0 EUR
METN52002577
pH sensor, InPro3253/SG/120/PT1000
Elektroden pH-Elektroden
High performance pH and ORP measurement.
  • Optimized for steam sterilization
  • Low maintenance for your operation
  • Sterilizable and autoclavable
The InPro 3253SG 120/PT1000 is a pre-pressurized, liquid filled, low maintenance pH and ORP (Redox) sensor combined with a PT1000 temperature sensor for in-line measurement in demanding applications. This version is 120 mm long.
The steam-sterilizable pH glass membrane of the InPro 3253SG 120/PT1000 makes it optimal for processes where sterilization or autoclaving are required.
This pH and ORP sensor is designed for low maintenance measurements, from dirty industrial to hygienic and sterile environments.
This sensor provides fast and precise pH and ORP measurement, even after repeated autoclaving and sterilization cycles.
Platinum solution ground/redox metal.
Standards: CE; ATEX/IECEx: Ex ia IIC T6 / T5 / T4 / T3 Ga / Gb; EHEDG; FM: IS Cl. I, II, III, Div 1, GR ABCDEFG / T6; METTLER TOLEDO Quality Certificate; PED.