DNA (Deoxyribonucleic acid) (aus Lachshoden), beschallt
Lieferant: Cytiva
27-4565-01EA 498 EUR
27-4565-01
DNA (Deoxyribonucleic acid) (aus Lachshoden), beschallt
DNS (Desoxyribonukleinsäure)
Genomic DNA sheared by sonication for use in hybridization experiments
  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • Suitable for blocking non-specific binding sites in hybridization experiments to reduce background
Sonicated DNA from salmon testes.
Lagertemperatur: Gefrierschrank
MDL: MFCD00130922
CAS-Nummer: 9007-49-2