Mineralöl, weiß stabilisiert USP, J.T.Baker®

Lieferant: Avantor
Gefahr


Stabilisierung: Stabilisiert mit Vitamin E 10-100 ppm
Veränderungsmanagement-Kategorie= R

2705-01EA 57.01 EUR
2705-01
Mineralöl, weiß stabilisiert USP, J.T.Baker®
Mineralöl
Siedepunkt: 260 °C
Dichte: 0,85 g/cm³ (20 °C)
Flammpunkt: 135 °C (geschlossener Tiegel)
MDL: MFCD00131611
CAS-Nummer: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Acidity Passes Test
USP - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
USP - Limit of Sulfur Compounds Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.845 - 0.905
USP - Viscosity at 40 C, mm₂/s 34.5 - 150.0
USP - Readily Carbonizable Substances Passes Test
USP - Solid Paraffin Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR