Chicken blood, sterile
Lieferant: Rockland Immunochemicals
ROCKR302-0100EA 309 EUR
ROCKR302-0100 ROCKR302-0050
Chicken blood, sterile
Blut
Chicken whole blood aseptically collected. These cell suspensions are ideal for washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.