Digital thermo-hygrometer, KlimaLogg Pro
Lieferant: TFA Dostmann
DOST303039.ITEA 167 EUR
DOST303039.IT
Digital thermo-hygrometer, KlimaLogg Pro
Hygrometer Thermohygrometer, digital
This instrument is designed to measure temperature humidity and time.
  • Graph display and switching output
  • Up to 50000 measured values
  • Radio controlled clock with seconds, date, time and zone in 12 hours
  • Maximum/minimum values and current dew point of the selected channel
  • Alarm function that can be desactivated