Sample loops for Valco valves
Lieferant: COLLECTION PREFIX HPLC
HPLCSL100CWEA 0 EUR
HPLCSL100CW
Sample loops for Valco valves
Chromatographie-Zubehör Chromatographie-Probenschläuche
These stainless steel sample loops are designed for use with valco valve models CW6 and EC6W. Each loop has burr-free, polished ends and is passivated and flushed with reagent-grade methanol to ensure cleanliness.