Anti-Cystathionase Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB125210-100EA 816 EUR
ABCAAB125210-100
Anti-Cystathionase Rabbit Polyclonal Antibody
Type: Primary
Antigen: Cystathionase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR