Fibroblast growth medium 2
Lieferant: PromoCell
C-23120EA 231 EUR
C-23120 C-23220 C-23020 C-23225
Fibroblast growth medium 2
Zellkulturmedien
Fibroblast Growth Medium 2 is a low serum medium developed for adult human fibroblasts.
The medium is optimised for primary human cells but can also be used for rat, porcine and bovine fibroblasts as well as fibroblast cell lines.
Lieferumfang: Fibroblast Growth Medium 2 (ready to use) consists of a 500 ml bottle of basal medium and one vial of SupplementMix (growth supplements mixed together in one tube). Fibroblast Growth Medium 2 kit consists of a 500 ml bottle of basal medium and the SupplementPack (growth supplements come in separate vials).