Interlining Gun
Lieferant: Steinel
238-5660EA 53.4 EUR
238-5660
Interlining Gun
Heizgeräte Heißlufttrockner
Interlining gun with electronically controlled heating and high power - for professionals and amateurs .Simple operation due to mechanical feed device glue sticks and high power melting brought the gun into the category of first-class tool for users who prefer quality products.
  • Automatic self-adhesive keeps the temperature constant
  • Economical power consumption - 45 W in the operating phase and 16 W in the standby phase, during the initial heating of 200 W
  • High proizvoditelnsot - flow of about 16 grams of adhesive per minute
  • Short heating time about 7 minutes and Quick drying glue for 2 minutes