Gas filter cartridge filters for Ekastu masks
Lieferant: Klein Tools
111-3411EA 46 EUR
111-3411
Gas filter cartridge filters for Ekastu masks
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
These filters protect against organic gases and vapours with a boiling point >65 °C.
  • Inhalation resistance: at 30 L/min, constant flow max. 1,4 mbar; at 95 L/min, constant flow max. 5,6 mbar
  • Both filter openings are locked by water-vapour-proof cover caps
Filters can be used in connection with full masks (DIN EN 136) and half masks (DIN EN 140) with round thread connection (DIN EN 148-1).
Standards: DIN EN 14387