Surgical mask, Tie-on, type II, 1818
Lieferant: 3M
114-3440EA 82.6 EUR
MMMEM1818
Surgical mask, Tie-on, type II, 1818
These flat surgical masks with aluminium nose band and adjustable tie provides comfort, breathability and maximum protection.
  • Provides 98% bacterial filtration efficiency
  • Hypoallergenic to reduce chance of allergic reaction
  • Helps reduce potential exposure of the wearer to blood and body fluids
Standards: EN 14683:2005