Mask packages, H 68 series
Lieferant: Entegris
ENTEH6862550967C02EA 0 EUR
ENTEH6862550967C02
Mask packages, H 68 series
Wafer-Handling-Zubehör Wafer-Carrier
For masks or reticles with pellicles. Label for identication labels. Landing pads prevent tearing of pellicies.
  • Minimal contact
  • Easy to open and close
  • Easy removal
  • Seal security for protection
  • Compact and stackable
  • Roller hinge minimises particle generation