Digital timer with alarm
Lieferant: Moineau
MOIN437/2EEA 77 EUR
MOIN437/2E
Digital timer with alarm
Timer Timer, digital
This digital timer offers four measuring channels (1× count up, 3× count down).
  • Timing capacity: 20 hours (1/100 seconds)
  • Independent or simultaneous measurements
  • With clock
  • Three alarms, three memories
Lieferumfang: Supplied with stand, magnet and two LR44 batteries.