Ampicillin, HP Ampillicin™

Lieferant: G Bioscienes

GD-6060EA 397 EUR
GD-6060
Ampicillin, HP Ampillicin™
Ampicillin

HP Ampicillin™ is a stronger and more stable inhibitor of ß-lactamase than regular ampicillin and therefore supports only the growth of ampicillin resistant colonies even after prolonged incubation. Culture plates can be incubated for a longer period at 37 °C, eliminating the time constraint associated with normal ampicillin.


  • Reduces satellite colonies
  • Lyophilised format for improved stability
  • Reconstituted 1000X concentration
  • Suitable for 5 L media or agar


Lieferumfang: HP Ampicillin™ is supplied lyophilised. When reconstituted in 500 µl deionised water it produces a 1000X solution. Each kit contains 10 vials, suitable for a total volume of 5 L culture media or agar.

Formel: C₁₆H₁₉N₃O₄S
Molecular Weight: 349,4 g/mol
MDL: MFCD00005175
CAS-Nummer: 69-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR