Ferrules for gas chromatography
Lieferant: Restek
REST21060EA 60.5 EUR
REST21060 REST21061 REST20250 REST20251 REST20252 REST20253 REST20254 REST20210 REST21058 REST20200 REST20201 REST21059 REST20246 REST20202 REST20203 REST20248 REST24848 REST20204 REST20205 REST24849 REST20249 REST20206 REST20207 REST20208 REST20209 REST27070 REST20281 REST20240 REST22222 REST22214 REST20233 REST22213 REST22216 REST22215 REST21007 REST22218 REST20238 REST21008 REST22217 REST20239 REST22219 REST27062 REST22210 REST20230 REST22212 REST20275 REST20231 REST22211 REST20232 REST20222 REST21036 REST20223 REST21037 REST21038 REST20224 REST20225 REST20226 REST20227 REST20228 REST20229 REST20263 REST20220 REST20264 REST20221 REST20255 REST20211 REST20212 REST20257 REST20213 REST20258 REST20215 REST20217 REST20218 REST20219
Ferrules for gas chromatography
Fittings und Verbinder
Ferrules are available in Vespel®, graphite or Vespel®/graphite.
Vespel® ferrules: Ferrules are made up of 100% high-temperature polyimide. Stable to 350 °C, durable and leak tight.
Graphite ferrules: Ferrules are preconditioned to eliminate out-gassing. High purity, high density graphite. Smoother surface and cleaner edges than conventional graphite ferrules. Contain no binders that can off-gas or adsorb analytes. Stable to 450 °C.
Vespel®/graphite ferrules: VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability. VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules. Sealed with minimal torque, reusable and preferred for vacuum and high-pressure uses. Stable to 400 °C, recommended for mass spec transfer lines.