Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Lieferant: Thermo Scientific
12942.03EA
51.5
EUR
12942.03
12942.09
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
Expiration date provided