Silver lactate
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Lactic acid silver salt
20835.09
20835.18
20835.09EA
134
EUR
20835.09
20835.18
Silver lactate
Silver lactate
CAS No.: 128-00-7
Molecular Formula: C3H5AgO3
Formula Weight: 196.94
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to grey
Odour: Odourless
Melting Point: 120-122°
UN No.: UN3077
MDL No.: MFCD00043279
Formel:
C₃H₅AgO₃ Molecular Weight: 196,938 g/mol Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043279 CAS-Nummer: 15768-18-0 ADR: 9,III Merck Index: 14,8517 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...