PTFE, Tafeln, Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Scientific

45172.HBEA 169 EUR
45172.HB
PTFE, Tafeln, Dicke 3.18 mm (0.125 in)
PTFE
Formel: (CF₂CF₂)n
Lagertemperatur: Raumtemperatur
MDL: MFCD00084455
CAS-Nummer: 9002-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR