Platin ≥99,9% (Metall-Basis), amorphes Pulver APS 2.0-8.0 micron
Lieferant: Thermo Scientific
Synonyme: Platinium
47145.03 47145.06
47145.03EA 356 EUR
47145.03 47145.06
Platin ≥99,9% (Metall-Basis), amorphes Pulver APS 2.0-8.0 micron
Platinum
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: S.A. 0.5-1.5m2/g, Tap density 0.5-2.5g/cm3 Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Molecular Weight: 195,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011179
CAS-Nummer: 7440-06-4
EINECS: 231-116-1
ADR: 4.1,II