Kaliumpyroantimonat ≥94%

Lieferant: Thermo Scientific
Achtung

39524.30EA 406 EUR
39524.30 39524.18
Kaliumpyroantimonat ≥94%
Kaliumpyroantimonat
Formel: KSbO₃·3H₂O
Molecular Weight: 262,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011415
CAS-Nummer: 12208-13-8
EINECS: 235-387-7
UN: 1549
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR