cis-3-Chloroacrylic acid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cis-3-chloroacrylicacid

A14798.03 A14798.06
A14798.03EA 45 EUR
A14798.03 A14798.06
cis-3-Chloroacrylic acid ≥98%
cis-3-Chloroacrylic acid

Purity: 98+% CAS No.: 1609-93-4 Molecular Formula: C3H3ClO2 Formula Weight: 106.51 Storage Temperature: 2°C to 8°C Melting Point: 60-62° Beilstein Reference: 1720278 UN No.: UN3261 MDL No.: MFCD00004368

Formel: C₃H₃ClO₂
Molecular Weight: 106,51 g/mol
Schmelzpunkt: 60…62 °C
MDL: MFCD00004368
CAS-Nummer: 1609-93-4
EINECS: 216-545-4
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR