Ellagic acid, (∼12% H₂O) 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3,7,8-Tetrahydroxychromeno[5,4,3-cde]chromene-5,10-dione, 4,4',5,5',6,6'-Hexahydroxydiphenic acid 2,6,2',6'-dilactone
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.
Purity: 97%
CAS No.: 476-66-4
Molecular Formula: C14H6O8.xH2O
Formula Weight: 302.20(anhy)
Melting Point: 450° dec.
Beilstein Reference: 47549
MDL No.: MFCD00149494
Formel:
C₁₄H₆O₈·xH₂O Molecular Weight: 302,194 g/mol Schmelzpunkt: 450 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149494 CAS-Nummer: 476-66-4 EINECS: 207-508-3 Merck Index: 14,3547 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...