Aluminiumoxid aktiviert, neutral Brockmann Grade II

Lieferant: Thermo Scientific

A17141.0E A17141.36
A17141.0EEA 164 EUR
A17141.0E A17141.36
Aluminiumoxid aktiviert, neutral Brockmann Grade II
Aluminium oxide activated neutral

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Flash Point: None Odour: Odourless MDL No.: MFCD00003424

Formel: Al₂O₃
Molecular Weight: 101,96 g/mol
Dichte: 3,97
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR