Chlorodi(1-naphthyl)phosphine 95%
Lieferant: Thermo Scientific
|
Gefahr
|
H25893.06
H25893.03
H25893.06EA
482
EUR
H25893.06
H25893.03
Chlorodi(1-naphthyl)phosphine 95%
Chlorodi(1-naphthyl)phosphine
Purity: 95%
CAS No.: 36042-99-6
Molecular Formula: C20H14ClP
Formula Weight: 320.76
Melting Point: 154-157°
Boiling Point: 227-229°/0.6mm
UN No.: UN3261
MDL No.: MFCD09753469
Formel:
C₂₀H₁₄ClP Molecular Weight: 320,76 g/mol Siedepunkt: 227…229 °C (0,6 mmHg) Schmelzpunkt: 154…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753469 CAS-Nummer: 36042-99-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...