Triphenylsulphonium chloride 94%

Lieferant: Thermo Scientific
Achtung

H26059.03EA 83.9 EUR
H26059.03
Triphenylsulphonium chloride 94%
Triphenylsulphonium chloride
Formel: (C₆H₅)₃SCl
Molecular Weight: 298,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031716
CAS-Nummer: 4270-70-6
EINECS: 224-259-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR