6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one 96%

Lieferant: Thermo Scientific
Achtung

H62026.03EA 361 EUR
H62026.03
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one 96%
6-Bromo-7-fluoro-2,4-dihydro-1,4-benzoxazin-3-one
Formel: C₈H₅BrFNO₂
Molecular Weight: 246,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441932
CAS-Nummer: 355423-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR