Ethyl orange sodium salt

Lieferant: Thermo Scientific
Achtung

J62863.22EA 30.7 EUR
J62863.22
Ethyl orange sodium salt
Ethyl orange sodium salt

Applications: Indicator: pH 3.0 (red) to pH 4.5 (yellow)

Formel: C₁₆H₁₈N₃NaO₃S
Molecular Weight: 355,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007503
CAS-Nummer: 62758-12-7
EINECS: 263-716-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR