G418 Disulfat 50 mg/ml Lösung
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat
J63871.AB
J63871.AD
J63871.ABEA
119
EUR
J63871.AB
J63871.AD
G418 Disulfat 50 mg/ml Lösung
G418 disulphate
CAS No.: 108321-42-2
Molecular Formula: C20H40N4O10.2H2SO4
Formula Weight: 692.71
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Clear
MDL No.: MFCD00058314
Applications: Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths
Formel:
C₂₀H₄₄N₄O₁₈S₂ Molecular Weight: 692,72 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00058314 CAS-Nummer: 108321-42-2 |